missing translation for 'onlineSavingsMsg'
Learn More

Nuclear Membrane Marker Antibody (AE-5), DyLight 488, Novus Biologicals™

Product Code. 18398211 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18398211 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18398211 Supplier Novus Biologicals Supplier No. NBP311308G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Nuclear Membrane Marker Monoclonal antibody specifically detects Nuclear Membrane Marker in Human samples. It is validated for Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nuclear Membrane Marker
Applications Immunofluorescence
Classification Monoclonal
Clone AE-5
Conjugate DyLight 488
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Nuclei of myeloid leukemia biopsy cells
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.