missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nuclear Matrix Protein p84 Monoclonal Antibody (301CT40.5.1)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA537530

Product Code. 17157118

  • £466.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Nuclear Matrix Protein p84 Monoclonal Antibody for Western Blot

HPR1 is part of the TREX (transcription/export) complex, which includes TEX1 (MIM 606929), THO2 (MIM 300395), ALY (MIM 604171), and UAP56 (MIM 142560).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nuclear Matrix Protein p84
Monoclonal
0.25 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q96FV9
Thoc1
KLH conjugated synthetic peptide between 257-285 amino acids from the N-terminal region of human THOC1.
400 μL
Primary
Human
Antibody
IgG3 κ
Western Blot
301CT40.5.1
Unconjugated
Thoc1
3110002N20Rik; AW107452; HPR1; hTREX84; I79_014746; LRRGT00070; NMP-84; Nuclear matrix protein p84; P84; P84N5; THO complex 1; THO complex subunit 1; THO complex subunit 1-like protein; tho1; Thoc1
Mouse
Protein G
RUO
9984
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.