missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Nuclear Matrix Protein p84 Monoclonal Antibody (5E10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA123261
This item is not returnable.
View return policy
Description
Suggested positive controls are HeLa, Raji, and MOLT4.
HPR1 is part of the TREX (transcription/export) complex, which includes TEX1 (MIM 606929), THO2 (MIM 300395), ALY (MIM 604171), and UAP56 (MIM 142560).
Specifications
| Nuclear Matrix Protein p84 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7 | |
| P59924, Q8R3N6, Q96FV9 | |
| Thoc1 | |
| Amino acids 15-374 of human p84 expressed in E. coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey, Hamster | |
| Antibody | |
| IgG2b |
| ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 5E10 | |
| Unconjugated | |
| Thoc1 | |
| 3110002N20Rik; AW107452; HPR1; hTREX84; I79_014746; LRRGT00070; NMP-84; Nuclear matrix protein p84; P84; P84N5; THO complex 1; THO complex subunit 1; THO complex subunit 1-like protein; tho1; Thoc1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 100770844, 225160, 291797, 9984 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction