missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nuclear Fast Red Soln., enhanced stability

Product Code. 17887638
Click to view available options
Quantity:
500 mL
1000 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 17887638

Brand: Thermo Scientific Chemicals J61010.AU

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A histological counterstain. Nuclear Fast Red solution turns cell nuclei red and cytoplasm a pale pink.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nuclear Fast Red Soln.
Synonym Kernechtrot Solution
Packaging Plastic bottle
Quantity 1000 mL
Physical Form Liquid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.