missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nuclear Factor Erythroid Derived 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31540
This item is not returnable.
View return policy
Description
Nuclear Factor Erythroid Derived 2 Polyclonal antibody specifically detects Nuclear Factor Erythroid Derived 2 in Human samples. It is validated for Western Blot, ChIP assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation.
Specifications
| Nuclear Factor Erythroid Derived 2 | |
| Polyclonal | |
| Unconjugated | |
| 1XPBS, 1% BSA, 20% Glycerol (pH7). with 0.025% Proclin 300 | |
| NFE2 | |
| Recombinant protein encompassing a sequence within the center region of human NF-E2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 4778 | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, ChIP Assay | |
| 1.03mg/mL | |
| Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP) | |
| Leucine zipper protein NF-E2, NF-E2, nuclear factor (erythroid-derived 2), 45kD, nuclear factor (erythroid-derived 2), 45kDa, Nuclear factor, erythroid-derived 2 45 kDa subunit, p45, p45 NF-E2, transcription factor NF-E2 45 kDa subunit | |
| Rabbit | |
| 41 kDa | |
| 0.1 mL | |
| Primary | |
| Tested in human. Predicted cross-reactivity based on sequence identity: Monkey (97%). Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction