missing translation for 'onlineSavingsMsg'
Learn More

Nuclear Factor Erythroid Derived 2 Antibody (2C6), Novus Biologicals™

Product Code. 18336577 Shop All Bio Techne Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
0.1 mg
missing translation for 'unitSize'
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18336577 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 18336577 missing translation for 'mfr' Novus Biologicals missing translation for 'supplierNo' H00004778M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Nuclear Factor Erythroid Derived 2 Monoclonal antibody specifically detects Nuclear Factor Erythroid Derived 2 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nuclear Factor Erythroid Derived 2
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2C6
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH05044
Gene Alias Leucine zipper protein NF-E2, NF-E2, nuclear factor (erythroid-derived 2), 45kD, nuclear factor (erythroid-derived 2), 45kDa, Nuclear factor, erythroid-derived 2 45 kDa subunit, p45, p45 NF-E2, transcription factor NF-E2 45 kDa subunit
Host Species Mouse
Immunogen NFE2 (AAH05044, 1 a.a. ~ 373 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSPCPPQQSRNRVIQLSTSELGEMELTWQEIMSITELQGLNAPSEPSFEPQAPAPYLGPPPPTTYCPCSIHPDSGFPLPPPPYELPASTSHVPDPPYSYGNMAIPVSKPLSLSGLLSEPLQDPLALLDIGLPAGPPKPQEDPESDSGLSLNYSDAESLELEGTEAGRRRSEYVEMYPVEYPYSLMPNSLAHSNYTLPAAETPLALEPSSGPVRAKPTARGEAGSRDERRALAMKIPFPTDKIVNLPVDDFNELLARYPLTESQLALVRDIRRRGKNKVAAQNCRKRKLETIVQLERELERLTNERERLLRARGEADRTLEVMRQQLTELYRDIFQHLRDESGNSYSPEEYALQQAADGTIFLVPRGTKMEATD
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4778
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.