missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nuclear Factor Erythroid 2 Related Factor 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-87955
This item is not returnable.
View return policy
Description
Nuclear Factor Erythroid 2 Related Factor 1 Polyclonal specifically detects Nuclear Factor Erythroid 2 Related Factor 1 in Human samples. It is validated for Western Blot.
Specifications
| Nuclear Factor Erythroid 2 Related Factor 1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 4779 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| FLJ00380, HBZ17, LCR-F1, Locus control region-factor 1, NF-E2-related factor 1, NFE2-related factor 1, NRF1TCF11, nuclear factor (erythroid-derived 2)-like 1, nuclear factor erythroid 2-related factor 1, Nuclear factor, erythroid derived 2, like 1, TCF-11, Transcription factor 11, transcription factor 11 (basic leucine zipper type), Transcription factor HBZ17, Transcription factor LCR-F1 | |
| The immunogen is a synthetic peptide directed towards the middle region of human Nuclear Factor Erythroid 2 Related Factor 1 | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction