missing translation for 'onlineSavingsMsg'
Learn More

Nuclear Antigen Rabbit anti-Human, Mouse, Rat, Clone: NM2984R, Novus Biologicals™

Product Code. 18328211 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18328211 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18328211 Supplier Novus Biologicals Supplier No. NBP30730320UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Nuclear Antigen Monoclonal specifically detects Nuclear Antigen in Human, Mouse, Rat samples. It is validated for Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nuclear Antigen
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone NM2984R
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 0.25-0.5 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Host Species Rabbit
Immunogen Nuclei of HL60 cells
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.