missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Nuclear Pore Complex Monoclonal Antibody (39C7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA533097
This item is not returnable.
View return policy
Description
Nuclear Pore Complex Monoclonal Antibody for ICC/IF
The nuclear pore complex (NPC) serves as the sole bidirectional gateway of macromolecules in and out of the nucleus. Each NPC consists of ~1,000 protein subunits, termed nucleoporins, and totals to a molecular mass of ~110 Mda. NPCs maintain the integrity of the nuclear compartment by preventing macromolecules from freely diffusing in or out of the nucleus. (Annu Rev Biochem, 2019. PMID 30883195).
Specifications
| Nuclear Pore Complex | |
| Monoclonal | |
| 0.25 mg/mL | |
| tissue culture supernatant with 10mM sodium azide | |
| O08587, O08658, O75694, P11654, P17955, P35658, P37198, P37199, P49790, P49791, P49793, P52590, P52948, P57740, Q12769, Q4QQS8, Q63850, Q66HC5, Q68FY1, Q6PFD9, Q80U93, Q8BH74, Q8BJ71, Q8CEC0, Q8N1F7, Q8NFH5, Q8R0G9, Q8R480, Q8R4R6, Q8TEM1, Q8WUM0, Q92621, Q99567, Q99P88, Q9BW27, Q9JIH2, Q9QY81, Q9UKX7, Q9Z0W3 | |
| NUP107, NUP133, NUP153, NUP155, NUP160, NUP205, NUP210, NUP214, NUP35, NUP50, NUP62, NUP85, NUP88, NUP93, NUP98 | |
| Yeast nuclear preparations. | |
| RUO | |
| 103468, 10762, 113929, 116555, 117021, 129401, 170762, 18141, 18226, 19069, 218210, 227720, 23165, 23225, 23279, 234865, 23636, 25281, 25497, 269966, 287830, 291874, 292085, 295692, 296634, 311182, 362335, 445007, 4927, 4928, 54563, 55746, 57122, 58958, 59015, 65274, 69482, 70699, 71805, 79902, 8021, 81738, 9631, 9688, 9972 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 39C7 | |
| Unconjugated | |
| NUP155 | |
| 107 kDa nucleoporin; 133 kDa nucleoporin; 153 kDa nucleoporin; 155 kDa nucleoporin; 160 kDa nucleoporin; 160kDa; 1700030K07Rik; 205 kDa nucleoporin; 214 kDa nucleoporin; 2310006I24Rik; 2410008G02Rik; 2810011M03Rik; 35 kDa nucleoporin; 35kDa; 3830404O05Rik; 4732457F17; 50 kDa nucleoporin; 5330402E05Rik; 62 kDa nucleoporin; 85 kDa nucleoporin; 88 kDa nuclear pore complex protein; 88 kDa nucleoporin; 93 kDa nucleoporin; 96 kDa nucleoporin; 98 kDa nucleoporin; 9830001L10; AA414952; AA589433; ADAR2; ADIR2; AI413123; AI426861; AI790512; AI836801; AI849286; ATFB15; AU020188; AU045898; AV248391; AW541137; B130015D15Rik; BC039282; C76801; C7orf14; C88147; CAIN; CAN; CAN protein, putative oncogene; CBP-interacting protein 4; Cip4; D2H9S46E; D7Ertd649e; D930027M19Rik; FROUNT; gene trap locus 1-13 protein; gene trap locus-13; GLFG-repeat containing nucleoporin; gp190; GP210; Gtl1-13; Gtl13; Gtl-13; hNUP133; HNUP153; IBSN; karyoporin; KIAA0023; Kiaa0095; Kiaa0197; KIAA0225; KIAA0791; KIAA0906; mermaid; Mitotic phosphoprotein 44; mKIAA0197; mKIAA0225; mKIAA0791; Mp44; MP-44; N153; N155; NIC96; NO44; NP44; Np62; NPAP60; NPAP60L; NPHS11; NPHS12; NPHS13; Nuclear envelope pore membrane protein POM 210; nuclear pore associated protein; nuclear pore associated protein 60 kDa; nuclear pore complex 1; nuclear pore complex protein 88; nuclear pore complex protein hnup153; nuclear pore complex protein Nup107; Nuclear pore complex protein Nup133; nuclear pore complex protein Nup153; Nuclear pore complex protein Nup155; nuclear pore complex protein Nup160; Nuclear pore complex protein Nup205; nuclear pore complex protein Nup214; Nuclear pore complex protein Nup50; nuclear pore complex protein Nup53; Nuclear pore complex protein Nup85; nuclear pore complex protein Nup88; Nuclear pore complex protein Nup93; Nuclear pore complex protein Nup96; Nuclear pore complex protein Nup98; nuclear pore complex protein Nup98; nuclear pore complex protein Nup98-Nup96; nuclear pore complex protein Nup98-Nup96; nuclear pore glycoprotein 62; nuclear pore glycoprotein p62; Nuclear pore membrane glycoprotein 210; nuclear pore membrane protein 210; nuclear pore protein gp210; Nuclear pore-associated protein 60 kDa-like; nuclear pore-associated protein 60L; nucleoporin 107; nucleoporin 107kDa; nucleoporin 133; nucleoporin 133kD; nucleoporin 133kDa; nucleoporin 153; nucleoporin 153kD; nucleoporin 153kDa; nucleoporin 155; nucleoporin 155kD; nucleoporin 155kDa; nucleoporin 160; nucleoporin 160kD; nucleoporin 160kDa; nucleoporin 205; nucleoporin 205kDa; nucleoporin 210; nucleoporin 210; nuclear pore membrane glycoprotein 210; nuclear pore membrane protein 210; nucleoporin 210kDa; nucleoporin 214; nucleoporin 214kDa; nucleoporin 35; nucleoporin 35kDa; nucleoporin 50; nucleoporin 50 kDa; nucleoporin 50kD; nucleoporin 50kDa; nucleoporin 62; nucleoporin 62kD; nucleoporin 62kDa; nucleoporin 85; nucleoporin 85kDa; nucleoporin 88; nucleoporin 88kDa; nucleoporin 93; nucleoporin 93kDa; nucleoporin 98; nucleoporin 98kD; nucleoporin 98kDa; nucleoporin Nup107; Nucleoporin Nup133; nucleoporin Nup153; Nucleoporin Nup155; nucleoporin Nup160; Nucleoporin Nup205; nucleoporin Nup210; Nucleoporin Nup214; nucleoporin Nup35; Nucleoporin Nup50; nucleoporin NUP53; Nucleoporin Nup62; nucleoporin Nup75; Nucleoporin Nup84; nucleoporin NUP85; Nucleoporin Nup88; nucleoporin Nup93; Nucleoporin Nup96; Nucleoporin Nup98; nucleoprotein 50; Nucleoprotein 50kD; NUCZINK; Nup107; NUP120; Nup133; Nup153; NUP155; Nup160; NUP196; Nup205; NUP210; Nup214; Nup35; Nup50; NUP53; NUP62; NUP75; Nup84; NUP85; NUP88; Nup93; Nup96; Nup98; NUP98/PHF23 fusion 2 protein; Nup98-Nup96; Nupc1; p105; P140; p62; Pcnt1; pericentrin 1; pericentrin-1; POM210; pore membrane protein of 210 kDa; Prei2; preimplantation protein 2; PRO1146; Protein CAN; PSEC0245; RGD1304977; Rtp60; SNDI | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction