missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nuclear Antigen Antibody (NM106), Novus Biologicals™
Mouse Monoclonal Antibody
£211.00 - £373.00
Specifications
| Clone | NM106 |
|---|---|
| Applications | Immunohistochemistry (Paraffin) |
| Form | Purified |
| Isotype | IgG1 κ |
| Product Code | Brand | dimQuantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | dimQuantity | Price | Quantity & Availability | |||||
|
18738463
|
Novus Biologicals
NBP2-45273-0.02mg |
£211.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18779163
|
Novus Biologicals
NBP2-45273-0.1mg |
£373.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence
Nuclear Antigen Monoclonal specifically detects Nuclear Antigen in Human, Mouse, Rat, Porcine (Negative) samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| NM106 | |
| Immunohistochemistry (Paraffin) | |
| Purified | |
| IgG1 κ |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title