missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nuclear Antigen Antibody (235-1R), Biotin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-14102B
This item is not returnable.
View return policy
Description
Nuclear Antigen Monoclonal antibody specifically detects Nuclear Antigen in Human, Primate samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Specifications
| Nuclear Antigen | |
| Monoclonal | |
| Biotin | |
| Nuclei, Nucleus | |
| Nuclei of human myeloid leukemia biopsy cells | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG κ |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| 235-1R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Human, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction