missing translation for 'onlineSavingsMsg'
Learn More

Nuclear Antigen Antibody (235-1R), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18379861
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18379861 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18379861 Supplier Bio-Techne Supplier No. NBP314102AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Nuclear Antigen Monoclonal antibody specifically detects Nuclear Antigen in Human, Primate samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nuclear Antigen
Applications Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 235-1R
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias Nuclei, Nucleus
Host Species Rabbit
Immunogen Nuclei of human myeloid leukemia biopsy cells
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Primate
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.