missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nuclear Antigen Antibody (235-1), Novus Biologicals™
Mouse Monoclonal Antibody
£234.00 - £428.00
Specifications
| Antigen | Nuclear Antigen |
|---|---|
| Clone | 235-1 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 ug/mL, Immunoprecipitation 1000-2000 ug/500 ug protein lysate, Immunohistochemistry-Frozen, Flow (Intracellular) |
| Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Flow Cytometry (Intracellular Staining) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18448731
|
Novus Biologicals
NBP2-34342-0.1mg |
0.1 mg |
£428.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18443252
|
Novus Biologicals
NBP2-34342-0.02mg |
0.02 mg |
£234.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nuclear Antigen Monoclonal antibody specifically detects Nuclear Antigen in Human, Porcine, Primate, Chicken (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), Flow (Intracellular)Specifications
| Nuclear Antigen | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Flow Cytometry (Intracellular Staining) | |
| Unconjugated | |
| Mouse | |
| Human, Pig, Primate, Chicken (Negative), Mouse (Negative), Rat (Negative) | |
| Nuclei, Nucleus | |
| IgG1 κ | |
| Protein A or G purified |
| 235-1 | |
| Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 ug/mL, Immunoprecipitation 1000-2000 ug/500 ug protein lysate, Immunohistochemistry-Frozen, Flow (Intracellular) | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| Nuclei of human myeloid leukemia biopsy cells | |
| Primary | |
| Store at 4C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title