missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nuclear Antigen Antibody (235-1), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34525MFV610
This item is not returnable.
View return policy
Description
Nuclear Antigen Monoclonal antibody specifically detects Nuclear Antigen in Human, Porcine, Primate, Chicken (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Flow Cytometry, Flow (Intracellular)
Specifications
| Nuclear Antigen | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Human, Pig, Primate, Chicken (Negative), Mouse (Negative), Rat (Negative) | |
| Purified |
| Flow Cytometry | |
| 235-1 | |
| Flow Cytometry, Flow (Intracellular) | |
| Nuclei, Nucleus | |
| Nuclei of human myeloid leukemia biopsy cells | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction