missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUCKS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12023-2-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a nuclear protein that is highly conserved in vertebrates. The conserved regions of the protein contain several consensus phosphorylation sites for casein kinase II and cyclin-dependent kinases, two putative nuclear localization signals, and a basic DNA-binding domain. It is phosphorylated in vivo by Cdk1 during mitosis of the cell cycle.Specifications
| NUCKS1 | |
| Polyclonal | |
| Unconjugated | |
| NUCKS1 | |
| JC7, NUCKS, NUCKS1, P1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64709, 64710, 98415 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
| 0.44 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q80XU3, Q9H1E3 | |
| Nucks1 | |
| NUCKS1 Fusion Protein Ag2652 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction