missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUCKS1 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-74634JF549
This item is not returnable.
View return policy
Description
NUCKS1 Polyclonal antibody specifically detects NUCKS1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| NUCKS1 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| FLJ21480, FLJ32016, FLJ38536, NUCKSnuclear ubiquitous casein and cyclin-dependent kinases substrate, nuclear casein kinase and cyclin-dependent kinase substrate 1, nuclear ubiquitous casein kinase and cyclin-dependent kinase substrate, P1, potential LAG1 interactor | |
| The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human nuclear ubiquitous casein kinase and cyclin-dependent kinase substrate using the numbering given in entry NP_073568.2 (GeneID 64710). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 64710 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction