missing translation for 'onlineSavingsMsg'
Learn More

NUCKS1 Antibody (3G10), Novus Biologicals™

Product Code. 18411369 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411369 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411369 Supplier Novus Biologicals Supplier No. H00064710M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NUCKS1 Monoclonal antibody specifically detects NUCKS1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUCKS1
Applications Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Sandwich ELISA
Classification Monoclonal
Clone 3G10
Conjugate Unconjugated
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence 10 μg/mL, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_073568.2
Gene Alias FLJ21480, FLJ32016, FLJ38536, NUCKSnuclear ubiquitous casein and cyclin-dependent kinases substrate, nuclear casein kinase and cyclin-dependent kinase substrate 1, nuclear ubiquitous casein kinase and cyclin-dependent kinase substrate, P1, potential LAG1 interactor
Host Species Mouse
Immunogen NUCKS1 (NP_073568.2, 1 a.a. ∽ 243 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSRPVRNRKVVDYSQFQESDDADEDYGRDSGPPTKKIRSSPREAKNKRRSGKNSQEDSEDSEDKDVKTKKDDSHSAEDSEDEKEDHKNVRQQRQAASKAASKQREMLMEDVGSEEEQEEEDEAPFQEKDSGSDEDFLMEDDDDSDYGSSKKKNKKMVKKSKPERKEKKMPKPRLKATVTPSPVKGKGKVGRPTASKASKEKTPSPKEEDEEPESPPEKKTSTSPPPEKSGDEGSEDEAPSGED
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64710
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.