missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUCB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552551
This item is not returnable.
View return policy
Description
Immunogen sequence: MSEVDTNKDR LVTLEEFLKA TEKKEFLEPD SWETLDQQQF FTEEELKEYE NIIALQENEL KKKADELQKQ KEELQRQHDQ LEAQKLEYHQ VIQQMEQKKL QQGIPPSGPA GELKFEPR Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.
NUCB2 was initially identified as calcium-binding protein EF hand motif-containing protein that could bind with Necdin, a growth suppressor expressed primarily in postmitotic neurons. NUC2 is a secreted protein that is cleaved into three major peptide products, Nesfatin-1, Nesfatin-2, and Nesfatin-3, of which Nesfatin-1 has been found to cause the suppression of food intake in a leptin-independent manner. Other studies have suggested that NUCB2/Nesfatin-1 may also play roles in energy homeostasis and closely related neuroendocrine functions.
Specifications
| NUCB2 | |
| Polyclonal | |
| Unconjugated | |
| NUCB2 | |
| AI607786; Calnuc; DNA-binding protein NEFA; epididymis secretory protein Li 109; Gastric cancer antigen Zg4; HEL-S-109; LOW QUALITY PROTEIN: nucleobindin-2; NEFA; nesfatin 1; nesfatin-1; Nucb2; nucleobindin 2; nucleobindin-2; Nucleobindin-2/Nesfatin-1; nucleobindin-2-like protein; nucleobinding 2; p54; prepronefastin; prepronesfatin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4925 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P80303 | |
| NUCB2 | |
| Recombinant protein corresponding to Human NUCB2. Recombinant protein control fragment (Product #RP-88817). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction