Learn More
Invitrogen™ NUCB2 Monoclonal Antibody (K70044_2H5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555886
Description
Sequence of this protein is as follows: KTKVQNIHPV ESAKIEPPDT GLYYDEYLKQ VIDVLETDKH FREKLQKADI EEIKSGRLSK ELDLVSHHVR TKLDELKRQE VGRLRMLIKA KLDSLQDIGM DHQALLKQFD HLNHLNPDKF ESTDLDMLIK AATSDLEHYD KTRHEEFKKY EMMKEHERRE YLKTLNEEKR KEEESKFEEM KKKHENHPKV NHPGSKDQLK EVWEETDGLD PNDFDPKTFF KLHDVNSDGF LDEQELEALF TKELEKVYDP KNEEDDMVEM EEERLRMREH VMNEVDTNKD RLVTLEEFLK ATEKKEFLEP DSWETLDQQQ FFTEEELKEY ENIIALQENE LKKKADELQK QKEELQRQHD QLEAQKLEYH QVIQQMEQKK LQQGIPPSGP
NUCB2 was initially identified as calcium-binding protein EF hand motif-containing protein that could bind with Necdin, a growth suppressor expressed primarily in postmitotic neurons. NUC2 is a secreted protein that is cleaved into three major peptide products, Nesfatin-1, Nesfatin-2, and Nesfatin-3, of which Nesfatin-1 has been found to cause the suppression of food intake in a leptin-independent manner. Other studies have suggested that NUCB2/Nesfatin-1 may also play roles in energy homeostasis and closely related neuroendocrine functions.
Specifications
| NUCB2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P80303 | |
| NUCB2 | |
| Recombinant human NUCB2 (immunogen range is 31-410). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| K70044_2H5 | |
| Unconjugated | |
| NUCB2 | |
| AI607786; Calnuc; DNA-binding protein NEFA; epididymis secretory protein Li 109; Gastric cancer antigen Zg4; HEL-S-109; LOW QUALITY PROTEIN: nucleobindin-2; NEFA; nesfatin 1; nesfatin-1; Nucb2; nucleobindin 2; nucleobindin-2; Nucleobindin-2/Nesfatin-1; nucleobindin-2-like protein; nucleobinding 2; p54; prepronefastin; prepronesfatin | |
| Mouse | |
| Protein A | |
| RUO | |
| 4925 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.