missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUBPL Monoclonal Antibody (OTI5D5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525734
This item is not returnable.
View return policy
Description
NUBPL Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Required for the assembly of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I). May deliver of one or more Fe-S clusters to complex I subunits.
Specifications
| NUBPL | |
| Monoclonal | |
| 1.1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8TB37 | |
| Nubpl | |
| Human recombinant protein fragment corresponding to amino acids 1-250 of NUBPL produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI5D5 | |
| Unconjugated | |
| Nubpl | |
| 2410170E07Rik; C14orf127; huInd1; IND1; IND1 homolog; Iron-sulfur protein NUBPL; iron-sulfur protein required for NADH dehydrogenase; NUBPL; nucleotide binding protein like; nucleotide binding protein-like; nucleotide-binding protein-like; RGD1307232 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 80224 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction