missing translation for 'onlineSavingsMsg'
Learn More

NUBP1 Antibody, Novus Biologicals™

Product Code. 18324648 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18324648 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18324648 Supplier Novus Biologicals Supplier No. H00004682B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

NUBP1 Polyclonal antibody specifically detects NUBP1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUBP1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation PBS (pH 7.4)
Gene Accession No. NP_002475.2
Gene Alias cytosolic Fe-S cluster assembly factor NUBP1, MGC117406, MGC130053, NBP 1, NBP1, NBPMGC130052, nucleotide binding protein (e.coli MinD like), nucleotide binding protein 1 (E.coli MinD like), nucleotide binding protein 1 (MinD homolog, E. coli), Nucleotide-binding protein 1
Host Species Mouse
Immunogen NUBP1 (NP_002475.2, 1 a.a. - 320 a.a.) full-length human protein. MEEVPHDCPGADSAQAGRGASCQGCPNQRLCASGAGATPDTAIEEIKEKMKTVKHKILVLSGKGGVGKSTFSAHLAHGLAEDENTQIALLDIDICGPSIPKIMGLEGEQVHQSGSGWSPVYVEDNLGVMSVGFLLSSPDDAVIWRGPKKNGMIKQFLRDVDWGEVDYLIVDTPPGTSDEHLSVVRYLATAHIDGAVIITTPQEVSLQDVRKEINFCRKVKLPIIGVVENMSGFICPKCKKESQIFPPTTGGAELMCQDLEVPLLGRVPLDPLIGKNCDKGQSFFIDAPDSPATLAYRSIIQRIQEFCNLHQSKEENLISS
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 4682
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.