missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUB1/NYREN18 Antibody (OTI4H2), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73084APC
This item is not returnable.
View return policy
Description
NUB1/NYREN18 Monoclonal specifically detects NUB1/NYREN18 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry.
Specifications
| NUB1/NYREN18 | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| NUB1 | |
| Recombinant protein fragment corresponding to amino acids 1-326 of human NUB1/NYREN18 (NP_057202) produced in E.coli. | |
| 0.1 mL | |
| 51667.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI4H2 | |
| Flow Cytometry | |
| BS4, NEDD8 ultimate buster-1, negative regulator of ubiquitin-like proteins 1NUB1L, NY-REN-18 antigen, NYREN18NEDD8 ultimate buster 1, Renal carcinoma antigen NY-REN-18 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction