missing translation for 'onlineSavingsMsg'
Learn More

NUB1/NYREN18 Antibody (OTI2G1), DyLight 594, Novus Biologicals™

Product Code. 18411274 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411274 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411274 Supplier Novus Biologicals Supplier No. NBP273082DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NUB1/NYREN18 Monoclonal antibody specifically detects NUB1/NYREN18 in Human, Mouse, Rat, Canine samples. It is validated for Flow Cytometry, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUB1/NYREN18
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI2G1
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias BS4, NEDD8 μgtimate buster-1, negative regulator of ubiquitin-like proteins 1NUB1L, NY-REN-18 antigen, NYREN18NEDD8 μgtimate buster 1, Renal carcinoma antigen NY-REN-18
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-326 of human NUB1/NYREN18 (NP_057202) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51667
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.