Learn More
Invitrogen™ Human NUAK2, GST Tag Recombinant Protein
NUAK2 or SNF1/AMP kinase-related kinase (SNARK) is a member of the NUAK family of SNF1-like kinase 2. Recombinant full-length human NUAK2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Brand: Invitrogen™ PV6378
Code : NEW
Additional Details : Weight : 0.01000kg
Description
NUAK2 is activated by muscle contraction and is a unique mediator of contraction-stimulated glucose transport in skeletal muscle (1). NUAK2 is involved in cellular stress responses linked to obesity and type 2 diabetes. NUAK2 shows kinase activity against a synthetic test peptide and the activity in keratinocytes is increased by AMP and 5-amino-4-imidazolecarboxamide riboside, implying that AMPK kinase-dependent pathway can activate NAUK2. Glucose deprivation also increases NAUK2 activity in baby hamster kidney fibroblasts.
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Specifications
Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation | |
96.3kDa | |
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. | |
Human | |
GST Tag |
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.25mM DTT, 10mM glutathione, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol | |
1mg | |
SNARK, FLJ90349, DKFZP434J037, DKFZp686F01113 | |
Recombinant |