missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUAK2 Polyclonal Antibody

NUAK2 Polyclonal Antibody
Brand: Invitrogen PA588040
This item is not returnable.
View return policy
Description
NUAK2 Polyclonal Antibody for Western Blot
Specifications
| NUAK2 | |
| Polyclonal | |
| Unconjugated | |
| Nuak2 | |
| 1200013B22Rik; EMBL:AAH81899.1}; mKIAA0537; NUAK family kinase 2; NUAK family SNF1-like kinase 2; NUAK family, SNF1-like kinase, 2; NUAK2; nuak2 {ECO:0000312; Omphalocele kinase 2; Omphk2; SNARK; SNF1/AMP activated protein kinase; SNF1/AMP kinase-related kinase; SNF1/AMP-activated protein kinase; UV126 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 289419, 81788 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.61 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q66HE5, Q9H093 | |
| Nuak2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 479-628 of human NUAK2 (NP_1122143). | |
| 100 μL | |
| Primary | |
| Human, Rat, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction