missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUAK2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94277-0.1ml
This item is not returnable.
View return policy
Description
NUAK2 Polyclonal antibody specifically detects NUAK2 in Rat samples. It is validated for Western Blot
Specifications
| NUAK2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DKFZp434J037, EC 2.7.11, EC 2.7.11.1, FLJ90349, NUAK family SNF1-like kinase 2, NUAK family, SNF1-like kinase, 2, SNARKDKFZp686F01113, SNF1/AMP activated protein kinase, SNF1/AMP kinase-related kinase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 479-628 of human NUAK2 (NP_112214.3). VSGDPKEQKPPQASGLLLHRKGILKLNGKFSQTALELAAPTTFGSLDELAPPRPLARASRPSGAVSEDSILSSESFDQLDLPERLPEPPLRGCVSVDNLTGLEEPPSEGPGSCLRRWRQDPLGDSCFSLTDCQEVTATYRQALRVCSKLT | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 81788 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Dela en innehållskorrigering