missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human NUAK1 (ARK5), His Tag Recombinant Protein
Human NUAK1 (ARK5), His Tag Recombinant Protein
Brand: Invitrogen™ PR7569A
Code : NEW
Additional Details : Weight : 0.01000kg
Description
The overexpression of the NUAK1 (ARK5) kinase has been associated with tumor invasion and metastasis (colorectal neoplasm).
Specifications
NP_055655 | |
78.3 kDa | |
NUAK1 (ARK5) | |
Human | |
His Tag | |
Ser⁄Thr Kinases: CAMK Group | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |
Kinase Assay | |
100μg | |
CAMKL Kinase Family | |
Recombinant | |
Storage Buffer: 0 mM Tris pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% TritonX-100, 2 mM DTT, 50% Glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |