missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human NUAK1 (ARK5), His Tag Recombinant Protein
Human NUAK1 (ARK5), His Tag Recombinant Protein
Brand: Invitrogen™ PV4127
Code : 50
Additional Details : Weight : 0.05250kg
Description
NUAK1 (ARK5) is a serine⁄threonine kinase that plays a role in many cellular functions. The overexpression of this kinase has been associated with tumor invasion and metastasis (colorectal neoplasm).Specifications
NP_055655 | |
4790, 9891 | |
10 μg | |
CAMKL Kinase Family | |
Recombinant | |
Storage Buffer: 0 mM Tris pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% TritonX-100, 2 mM DTT, 50% Glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: CAMK Group | |
Omnia™ Bulk Substrate | |
NUAK family, SNF1-like kinase, 1 | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
Kinase Assay | |
78.3 kDa | |
NUAK1 (ARK5) | |
Human | |
His Tag | |
Biochemical binding/kinetic | |
Dry Ice | |
NUAK1 | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |