missing translation for 'onlineSavingsMsg'
Learn More

NU6300, MedChemExpress

Product Code. 16497638
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497638 10 mg 10mg
16477546 100 mg 100mg
16413657 50 mg 50mg
16470108 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16497638 Supplier Medchem Express Supplier No. HY18930
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress NU6300 is a covalent, irreversible and ATP-competitive CDK2 inhibitor with an IC50 value of 0.16 μM. NU6300 can be used for the research of eukaryotic cell cycle- and transcription-related.

NU6300 is a covalent, irreversible and ATP-competitive CDK2 inhibitor with an IC50 value of 0.16 μM. NU6300 can be used for the research of eukaryotic cell cycle- and transcription-related.

Specifications

Chemical Name or Material NU6300
CAS 2070015-09-5
Color White
Purity Grade Notes Research
Molecular Formula C20H23N5O3S
Quantity 10 mg
Solubility Information DMSO : ≥ 32 mg/mL (77.39 mM)
SMILES O=S(C(C=C1)=CC=C1NC2=NC(OCC3CCCCC3)=C4C(NC=N4)=N2)(C=C)=O
Molecular Weight (g/mol) 413.49
Formula Weight 413.49
Percent Purity 96.34%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.