missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NU 7026

Description
ATP-competitive inhibitor of DNA-dependent protein kinase (DNA-PK). Displays selectivity over other PIKK family enzymes (IC50 values are 0.23, 13.0, > 100 and > 100 μM for DNA-PK, PI3K, ATM and ATR respectively). Radiosensitizes both proliferating and quiescent mouse embryonic fibroblast cells to IR and inhibits DSB repair.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 281.31 |
| Purity | >98% |
| Molecular Weight (g/mol) | 281.311 |
| Chemical Name or Material | NU 7026 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?