missing translation for 'onlineSavingsMsg'
Learn More

NU 7026, MedChemExpress

Product Code. 16442847
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16442847

Brand: Medchem Express HY15719

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NU 7026 (LY293646) is a novel specific DNA-PK inhibitor with IC50 of 0.23 μM, also inhibits PI3K with IC50 of 13 μM.

NU 7026 (LY293646) is a novel specific DNA-PK inhibitor with IC50 of 0.23 μM, also inhibits PI3K with IC50 of 13 μM.

Specifications

Chemical Name or Material NU 7026
CAS 154447-35-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H15NO3
Quantity 50 mg
Synonym LY293646
Solubility Information DMSO : 2.5 mg/mL (8.89 mM; Need ultrasonic)
SMILES O=C1C2=CC=C3C=CC=CC3=C2OC(N4CCOCC4)=C1
Molecular Weight (g/mol) 281.31
Formula Weight 281.31
Percent Purity 99.43%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.