missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NTSR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534028
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla (100%) Monkey (94%) Marmoset, Panda, Dog, Horse (88%).
TFIP11 is a nuclear speckle-localized protein that may play a role in spliceosome disassembly in Cajal bodies (Stanek et al., 2008 [PubMed 18367544]).
Specifications
| NTSR1 | |
| Polyclonal | |
| Unconjugated | |
| NTSR1 | |
| bK445C9.6; FLJ22086; High-affinity levocabastine-insensitive neurotensin receptor; hNtr1; neurotensin receptor 1; neurotensin receptor 1 (high affinity); neurotensin receptor type 1; NT receptor-1; NT1 receptor; NT-1r; NTR; NTR1; NT-R-1; NTR-1; NTRH; NTRR; NTS1; NTS-1; Ntsr; Ntsr1; RNTR1; Spp382; TIP39 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4923 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| PBS with 0.1% sodium azide | |
| P30989 | |
| NTSR1 | |
| Synthetic 17 amino acid peptide from 3rd cytoplasmic domain of human NTSR1. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction