missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NTSR1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15382431
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15382431 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15382431 Supplier Invitrogen™ Supplier No. PA534027

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey (100%) Elephant (94%) Marmoset (89%) Hamster (83%).

TFIP11 is a nuclear speckle-localized protein that may play a role in spliceosome disassembly in Cajal bodies (Stanek et al., 2008 [PubMed 18367544]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NTSR1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene NTSR1
Gene Accession No. P30989
Gene Alias bK445C9.6; FLJ22086; High-affinity levocabastine-insensitive neurotensin receptor; hNtr1; neurotensin receptor 1; neurotensin receptor 1 (high affinity); neurotensin receptor type 1; NT receptor-1; NT1 receptor; NT-1r; NTR; NTR1; NT-R-1; NTR-1; NTRH; NTRR; NTS1; NTS-1; Ntsr; Ntsr1; RNTR1; Spp382; TIP39
Gene Symbols NTSR1
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from N-terminal extracellular domain of human NTSR1.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4923
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.