missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NTS2/NTSR2 Antibody [CoraFluor™ 1], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB100-56472CL1
This item is not returnable.
View return policy
Description
NTS2/NTSR2 Polyclonal antibody specifically detects NTS2/NTSR2 in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| NTS2/NTSR2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| neurotensin receptor 2, neurotensin receptor type 2, neurotensin receptor, type 2, NT-R-2, NTR2Levocabastine-sensitive neurotensin receptor | |
| A portion of amino acids 150-200 of human Neurotensin receptor 2 was used as the immunogen. | |
| 0.1 mL | |
| GPCR | |
| 23620 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu