missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NTS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103974
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NTS.
NTS is a common precursor for two peptides, neuromedin N and neurotensin. Neurotensin is a secreted tridecapeptide, which is widely distributed throughout the central nervous system, and may function as a neurotransmitter or a neuromodulator. It may be involved in dopamine-associated pathophysiological events, in the maintenance of gut structure and function, and in the regulation of fat metabolism. Neurotensin also exhibits antimicrobial activity against bacteria and fungi. Tissue-specific processing may lead to the formation in some tissues of larger forms of neuromedin N and neurotensin. The large forms may represent more stable peptides that are also biologically active.
Specifications
| NTS | |
| Polyclonal | |
| Unconjugated | |
| NTS | |
| 5033428E16Rik; Large neuromedin N; Neuromedin N; neurotensin; Neurotensin/neuromedin N; NmN; NMN-125; NN; NT; NT/N; NTS; NTS1; pro-neurotensin/neuromedin; Tail peptide | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 299757, 4922, 67405 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P20068, P30990, Q9D3P9 | |
| NTS | |
| A synthesized peptide derived from human NTS(Accession P30990), corresponding to amino acid residues G130-Y170. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction