missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NTRK2 Mouse anti-Human, Clone:10B6C4, Abnova™
Mouse monoclonal antibody raised against recombinant human NTRK2.
Brand: Abnova MAB17289.100ug
This item is not returnable.
View return policy
Description
This gene encodes a member of the neurotrophic tyrosine receptor kinase (NTRK) family. This kinase is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. Signalling through this kinase leads to cell differentiation. Mutations in this gene have been associated with obesity and mood disorders. Alternate transcriptional splice variants encoding different isoforms have been found for this gene. [provided by RefSeq]Specifications
| NTRK2 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. |
|
| NTRK2 | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 10B6C4 | |
| neurotrophic tyrosine kinase, receptor, type 2 | |
| In PBS (0.05% sodium azide) | |
| GP145-TrkB, TRKB | |
| Recombinant protein corresponding to amino acids 207-339 of of human NTRK2 from E. coli. | |
| 4915 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction