missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NTRC 824

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
NTRC 824 is a potent and selective non-peptide neurotensin (NTS) type-2 receptor antagonist (IC50 = 38 nM). Exhibits >150-fold selectivity for NTS2 over NTS1.
Specifications
Specifications
| Chemical Name or Material | N-[2-[5-[[(4-Methylphenyl)sulfonyl]amino]-3-(2,2,2-trifluoroacetyl)-1H-indol-1-yl]acetyl]-L-leucine |
| CAS | 1623002-61-8 |
| Quantity | 10 mg |
| Target | Neurotensin Receptor Antagonists |
| Molecular Formula | C25H26F3N3O6S |
| Formula Weight | 553.55 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction