missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NTNG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5107199
This item is not returnable.
View return policy
Description
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
Netrin G1 (NTNG1) belongs to a conserved family of proteins that act as axon guidance cues during vertebrate nervous system development (Nakashiba et al., 2000).
Specifications
| NTNG1 | |
| Polyclonal | |
| Unconjugated | |
| NTNG1 | |
| A930010C08Rik; AI853992; axon guidance molecule; Kiaa0976; laminet 1; laminet-1; LMNT1; netrin G1; netrin G1f; netrin-G1; Ntng1; RGD1563465; UNQ571/PRO1133; YLSR571 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 22854 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q9Y2I2 | |
| NTNG1 | |
| Synthetic peptide directed towards the C-terminal region of HUMAN NTNG1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction