missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NTNG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530447
This item is not returnable.
View return policy
Description
Recommended positive controls: HeLa, HepG2, HCT116, mouse brain. Predicted reactivity: Mouse (97%), Rat (98%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Netrin G1 (NTNG1) belongs to a conserved family of proteins that act as axon guidance cues during vertebrate nervous system development (Nakashiba et al., 2000).
Specifications
| NTNG1 | |
| Polyclonal | |
| Unconjugated | |
| NTNG1 | |
| A930010C08Rik; AI853992; axon guidance molecule; Kiaa0976; laminet 1; laminet-1; LMNT1; netrin G1; netrin G1f; netrin-G1; Ntng1; RGD1563465; UNQ571/PRO1133; YLSR571 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22854, 80883 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.87 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8R4G0, Q9Y2I2 | |
| NTNG1 | |
| Recombinant protein encompassing a sequence within the center region of human Netrin G1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction