missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NTNG1 Monoclonal Antibody (1010035)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA562077
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Netrin G1 (NTNG1) belongs to a conserved family of proteins that act as axon guidance cues during vertebrate nervous system development (Nakashiba et al., 2000).
Specifications
| NTNG1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9Y2I2 | |
| NTNG1 | |
| Human embryonic kidney cell, HEK293-derived human Netrin-G1a, His29-Gly503. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| 1010035 | |
| Unconjugated | |
| NTNG1 | |
| A930010C08Rik; AI853992; axon guidance molecule; Kiaa0976; laminet 1; laminet-1; LMNT1; netrin G1; netrin G1f; netrin-G1; Ntng1; RGD1563465; UNQ571/PRO1133; YLSR571 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 22854 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction