missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
NTNG1 Monoclonal antibody specifically detects NTNG1 in Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | NTNG1 |
| Applications | Western Blot |
| Immunogen | Mouse recombinant NTNG1 |
| Research Discipline | Neuroscience |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?