missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NTN4 Monoclonal Antibody (299612)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA560926
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
NTN4 contains 3 laminin EGF-like domains, 1 laminin N-terminal domain and 1 NTR domain. NTN4 may play an important role in neural, kidney and vascular development. It promotes neurite elongation from olfactory bulb explants. NTN4 belongs to a family of proteins related to laminins (see LAMA1, MIM 150320) Koch et al. (2000) [PubMed 11038171].[supplied by OMIM].
Specifications
| NTN4 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9HB63, Q9JI33 | |
| NTN4 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Netrin-4, Gly19-Val628. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 299612 | |
| Unconjugated | |
| NTN4 | |
| Beta-netrin; CDY; CDY1A; FLJ23180; FLJ77185; hepar-derived netrin-like protein; netrin 4; netrin 4; netrin-4; netrin-4; Ntn4; PRO3091; RGD1565947 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 57764, 59277 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction