missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NTM Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14373-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the IgLON (LAMP, OBCAM, Ntm) family of immunoglobulin (Ig) domain-containing glycosylphosphatidylinositol (GPI)-anchored cell adhesion molecules. The encoded protein may promote neurite outgrowth and adhesion via a homophilic mechanism. This gene is closely linked to a related family member, opioid binding protein/cell adhesion molecule-like (OPCML), on chromosome 11. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| NTM | |
| Polyclonal | |
| Unconjugated | |
| NTM | |
| HNT, IgLON family member 2, IGLON2, neurotrimin, NT, NTM, NTRI | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 235106, 50863 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q99PJ0, Q9P121 | |
| NTM | |
| NTM Fusion Protein Ag5746 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction