missing translation for 'onlineSavingsMsg'
Learn More
Learn More
nti-NHP CD45 Mouse anti-Rhesus, Cynomolgus, Baboon, BUV496, Clone: D058-1283, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 741185
This item is not returnable.
View return policy
Description
nti-NHP CD45 Monoclonal antibody specifically detects nti-NHP CD45 in Rhesus, Cynomolgus, Baboon samples. It is validated for Flow Cytometry
Specifications
| nti-NHP CD45 | |
| Monoclonal | |
| Brilliant Ultraviolet 496 | |
| Pan Leukocyte, NHP-specific; PTPRC; LCA; L-CA; Leukocyte Common Ag | |
| Rhesus peripheral whole blood | |
| RUO | |
| Rhesus Monkey, Cynomolgus, Baboon | |
| IgG1 κ |
| Flow Cytometry | |
| D058-1283 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV496 under optimal conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction