missing translation for 'onlineSavingsMsg'
Learn More
Learn More
nti-Integrin αvβ5 Mouse anti-Human, BUV496, Clone: ALULA, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 750002
This item is not returnable.
View return policy
Description
nti-Integrin αvβ5 Monoclonal antibody specifically detects nti-Integrin αvβ5 in Human samples. It is validated for Flow Cytometry
Specifications
| nti-Integrin αvβ5 | |
| Monoclonal | |
| Brilliant Ultraviolet 496 | |
| Integrin alpha v beta 5; αVβ5; CD51/β5 | |
| Mouse fibroblasts expressing αvβ5 | |
| RUO | |
| Human | |
| IgG2b κ |
| Flow Cytometry | |
| ALULA | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV496 under optimal conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction