missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NTF3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534025
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Mouse, Rat, Hamster, Elephant, Panda, Dog, Bovine, Opossum (100%) Turkey (93%).
The protein encoded by this gene is a member of the neurotrophin family, that controls survival and differentiation of mammalian neurons. This protein is closely related to both nerve growth factor and brain-derived neurotrophic factor. It may be involved in the maintenance of the adult nervous system, and may affect development of neurons in the embryo when it is expressed in human placenta. NTF3-deficient mice generated by gene targeting display severe movement defects of the limbs. The mature peptide of this protein is identical in all mammals examined including human, pig, rat and mouse.
Specifications
| NTF3 | |
| Polyclonal | |
| Unconjugated | |
| Ntf3 | |
| AI316846; AI835689; HDNF; Nerve growth factor 2; neurotrophic factor; neurotrophin 3; neurotrophin-3; neurotrophin-3 (HDNF/NT-3); NGF2; NGF-2; NT3; NT-3; NTF3; Ntf-3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100774728, 18205, 486731, 4908, 532393, 81737 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P18280, P20181, P20783, Q08DT3 | |
| Ntf3 | |
| Synthetic 15 amino acid peptide from N-Terminus of human NTF3 / Neurotrophin 3. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey, Bovine, Hamster | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction