missing translation for 'onlineSavingsMsg'
Learn More

PeproTech NTF3 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody

£177.00 - £2745.00

Specifications

Antigen NTF3
Concentration 0.1-1.0 mg/mL
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Biotin
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17803163
View Documents
PeproTech
500-P82BT-25UG
25 μg
£177.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17813453
View Documents
PeproTech
500-P82BT-50UG
50 μg
£228.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17873503
View Documents
Invitrogen
500-P82BT-1MG
2 x 500μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
30054678
View Documents
PeproTech
500-P82BT-500UG
500 μg
£1466.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

AA Sequence of recombinant protein: MYAEHKSHRG EYSVCDSESL WVTDKSSAID IRGHQVTVLG EIKTGNSPVK QYFYETRCKE ARPVKNGCRG IDDKHWNSQC KTSQTYVRAL TSENNKLVGW RWIRIDTSCV CALSRKIGRT. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human NT-3. Anti-Human NT-3-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hNT-3 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human NT-3 (500-P82G) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hNT-3. Western Blot: To detect hNT-3 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hNT-3 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P82BT-1MG will be provided as 2 x 500 μg

The protein encoded by this gene is a member of the neurotrophin family, that controls survival and differentiation of mammalian neurons. This protein is closely related to both nerve growth factor and brain-derived neurotrophic factor. It may be involved in the maintenance of the adult nervous system, and may affect development of neurons in the embryo when it is expressed in human placenta. NTF3-deficient mice generated by gene targeting display severe movement defects of the limbs. The mature peptide of this protein is identical in all mammals examined including human, pig, rat and mouse.
Specifications

Specifications

NTF3
ELISA, Western Blot
Biotin
Rabbit
Human
P20783
4908
E.coli-derived, 27.2 kDa Recombinant Human NT-3.
Antigen affinity chromatography
Ntf3
0.1-1.0 mg/mL
Polyclonal
Lyophilized
RUO
PBS with no preservative
AI316846; AI835689; HDNF; Nerve growth factor 2; neurotrophic factor; neurotrophin 3; neurotrophin-3; neurotrophin-3 (HDNF/NT-3); NGF2; NGF-2; NT3; NT-3; NTF3; Ntf-3
Ntf3
Primary
-20°C
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.