missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NTF3 Monoclonal Antibody (OTI5C7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524892
This item is not returnable.
View return policy
Description
NTF3 Monoclonal Antibody for Western Blot, IHC (P)
The protein encoded by this gene is a member of the neurotrophin family, that controls survival and differentiation of mammalian neurons. This protein is closely related to both nerve growth factor and brain-derived neurotrophic factor. It may be involved in the maintenance of the adult nervous system, and may affect development of neurons in the embryo when it is expressed in human placenta. NTF3-deficient mice generated by gene targeting display severe movement defects of the limbs. The mature peptide of this protein is identical in all mammals examined including human, pig, rat and mouse.
Specifications
| NTF3 | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P20783 | |
| Ntf3 | |
| Recombinant protein expressed in E.coli corresponding to amino acids 139-257 of NT3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5C7 | |
| Unconjugated | |
| Ntf3 | |
| AI316846; AI835689; HDNF; Nerve growth factor 2; neurotrophic factor; neurotrophin 3; neurotrophin-3; neurotrophin-3 (HDNF/NT-3); NGF2; NGF-2; NT3; NT-3; NTF3; Ntf-3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4908 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction