missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NTF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15538-1-AP-150UL
This item is not returnable.
View return policy
Description
The MW of this protein is 14 kDa, and this antibody specifically recognizes the 14 kDa protein.
The protein encoded by this gene is a cytosolic factor that facilitates protein transport into the nucleus. It interacts with the nuclear pore complex glycoprotein p62. This encoded protein acts at a relative late stage of nuclear protein import, subsequent to the initial docking of nuclear import ligand at the nuclear envelope. It is thought to be part of a multicomponent system of cytosolic factors that assemble at the pore complex during nuclear import.Specifications
| NTF2 | |
| Polyclonal | |
| Unconjugated | |
| NUTF2 | |
| NTF 2, NTF2, nuclear transport factor 2, NUTF2, Placental protein 15, PP15 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10204, 291981, 68051 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P61970, P61971, P61972 | |
| Nutf2 | |
| NTF2 Fusion Protein Ag7879 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction