missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NTE Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA535939

Product Code. 15507411

  • £449.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: U87-MG, SK-N-SH, IMR-32, SK-N-AS, rat brain. Predicted reactivity: Mouse (99%), Chicken (94%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a phospholipase that deacetylates intracellular phosphatidylcholine to produce glycerophosphocholine. It is thought to function in neurite outgrowth and process elongation during neuronal differentiation. The protein is anchored to the cytoplasmic face of the endoplasmic reticulum in both neurons and non-neuronal cells. Mutations in this gene result in autosomal recessive spastic paraplegia, and the protein is the target for neurodegeneration induced by organophosphorus compounds and chemical warfare agents. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NTE
Polyclonal
Unconjugated
pnpla6
5'-NT; 5'-nucleotidase; 5'-nucleotidase ecto; 5'-nucleotidase, ecto (CD73); AI661849; BNHS; CALJA; CD73; E5NT; ecto-5'-nucleotidase; eN; eNT; iPLA2delta; LNMS; MSws; Neuropathy target esterase; NT; NT5; NT5E; NTE; NTEMND; OMCS; patatin like phospholipase domain containing 6; patatin-like phospholipase domain containing 6; patatin-like phospholipase domain-containing protein 6; PNPLA6; Purine 5-Prime-Nucleotidase; RGD1564611; SPG39; Swiss cheese; Swiss-cheese; sws; wu:fy20f06
Rabbit
Antigen affinity chromatography
RUO
10908, 360753, 50767
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.4 mg/mL
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Q3TRM4, Q8IY17
pnpla6
Recombinant protein encompassing a sequence within the C-terminus region of human NTE. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.